Tisk…

3-Phenylpropylboronic acid pinacol ester ≥97%

Dodavatel: Thermo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane, 3-Phenyl-1-propylboronic acid pinacol ester

H28985.06 H28985.03
H28985.06EA 9540 CZK
H28985.06 H28985.03
3-Phenylpropylboronic acid pinacol ester ≥97%
3-Phenylpropylboronic acid pinacol ester

Purity: 97% CAS No.: 329685-40-7 Molecular Formula: C15H23BO2 Formula Weight: 246.16 MDL No.: MFCD09953505

Vzorec: C₁₅H₂₃BO₂
MW: 246,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953505
CAS číslo: 329685-40-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR