Tisk…

(2-Bromo-4-methoxyphenyl)acetic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-(2-Bromo-4-methoxyphenyl)acetic acid, 2-Bromo-4-methoxyphenylacetic acid

H27859.14 H27859.03 H27859.06
H27859.14EA 9540 CZK
H27859.14 H27859.03 H27859.06
(2-Bromo-4-methoxyphenyl)acetic acid ≥97%
(2-Bromo-4-methoxyphenyl)acetic acid

Purity: 97% CAS No.: 66916-99-2 Molecular Formula: C9H9BrO3 Formula Weight: 245.07 Melting Point: 127-131° MDL No.: MFCD02664700

Vzorec: C₉H₉BrO₃
MW: 245.07
Bod tání: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD02664700
CAS číslo: 66916-99-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR