Tisk…

4-Hydroxyphenylboronic acid pinacol ester ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxybenzeneboronic acid pinacol ester

H27836.06 H27836.03
H27836.06EA 7250 CZK
H27836.06 H27836.03
4-Hydroxyphenylboronic acid pinacol ester ≥97%
4-Hydroxyphenylboronic acid pinacol ester

Purity: 97% CAS No.: 269409-70-3 Molecular Formula: C12H17BO3 Formula Weight: 220.07 Melting Point: 114-117° MDL No.: MFCD02093756

Vzorec: C₁₂H₁₇BO₃
MW: 220,08 g/mol
Bod tání: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD02093756
CAS číslo: 269409-70-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR