3-Cyano-4-pyridineboronic acid pinacol ester 95%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-Cyanopyridine-4-boronic acid, pinacol ester
H27814.MD
H27814.03
H27814.MDEA
5360
CZK
H27814.MD
H27814.03
3-Cyano-4-pyridineboronic acid pinacol ester 95%
3-Cyano-4-pyridineboronic acid pinacol ester
Purity: 95%
CAS No.: 878194-92-4
Molecular Formula: C12H15BN2O2
Formula Weight: 230.07
Melting Point: 97-99°
MDL No.: MFCD09037481
Vzorec:
C₁₂H₁₅BN₂O₂ Bod tání: 97…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD09037481 CAS číslo: 878194-92-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...