Tisk…

2,5-Dibromotoluene ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 1,4-Dibromo-2-methylbenzene

A19418.06 A19418.14
A19418.06EA 989 CZK
A19418.06 A19418.14
2,5-Dibromotoluene ≥98%
2,5-Dibromotoluene

Purity: 98% CAS No.: 615-59-8 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 110°(230°F) Refractive Index: 1.6020 Odour: Aromatic Melting Point: 5-6° Boiling Point: 106-108°/10mm Beilstein Reference: 1859123 MDL No.: MFCD00000090

Vzorec: C₇H₆Br₂
Bod varu: 106…108 °C (10 mmHg)
Bod tání: 5…6 °C
Hustota: 1,813
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000090
CAS číslo: 615-59-8
EINECS: 210-437-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR