Tisk…

3,4-Dibromotoluene ≥98%

Dodavatel: Thermo Fisher Scientific

A13351.06 A13351.14 A13351.22
A13351.06EA 848 CZK
A13351.06 A13351.14 A13351.22
3,4-Dibromotoluene ≥98%
3,4-Dibromotoluene

Purity: 98+% CAS No.: 60956-23-2 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Flash Point: 116°(241°F) Refractive Index: 1.6000 Melting Point: -10° Boiling Point: 251° Beilstein Reference: 1931706 MDL No.: MFCD00079744

Vzorec: C₇H₆Br₂
MDL Number: MFCD00079744
CAS číslo: 60956-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR