Tisk…

(3-Bromopropyl)benzene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Bromo-3-phenylpropane

A14050.0E A14050.22 A14050.36
A14050.0EEA 18020 CZK
A14050.0E A14050.22 A14050.36
(3-Bromopropyl)benzene ≥98%
(3-Bromopropyl)benzene

Purity: 98% CAS No.: 637-59-2 Molecular Formula: C6H5CH2CH2CH2Br Formula Weight: 199.09 Flash Point: 101°(213°F) Refractive Index: 1.5460 Boiling Point: 219-220° Beilstein Reference: 2205527 MDL No.: MFCD00000257

Vzorec: BrH₂CCH₂CH₂(C₆H₅)
MW: 199,09 g/mol
Bod varu: 219…220 °C
Hustota: 1,308
Bod vzplanutí: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000257
CAS číslo: 637-59-2
EINECS: 211-294-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR