Allylbenzene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Phenyl-2-propene, 3-Phenylpropene
M-Clarity™ quality level = MQ100
A29402-100ML
A29402-25ML
SIALA29402-100MLEA
6800
CZK
SIALA29402-100ML
SIALA29402-25ML
Allylbenzene, Sigma-Aldrich®
Allylbenzene
Vzorec:
(C₆H₅)CH₂CHCH₂ MW: 118,18 g/mol Bod varu: 157 °C (1013 hPa) Bod tání: < –40 °C Hustota: 0,89 g/cm³ (20 °C) Bod vzplanutí: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008651 CAS číslo: 300-57-2 EINECS: 206-095-7 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...