Tisk…
Scoops, FoodScoop
Dodavatel: BURKLE
5371-0100 5371-0200 5371-0350 5371-0500 5371-0250
231-0403EA 871 CZK
231-0403 231-0404 231-0406 231-0407 231-0405
Scoops, FoodScoop
Lopatky
Stainless steel V2A (1.4301), highly polished. Very stable hand scoop for various applications, particularly suitable for the food industry.
  • Conforms to European food law regulations
  • Robust design
  • Seamless join between scoop and hollow handle so has no crevices and edges