Benzen ≥99.5% for HPLC
Dodavatel: abcr
|
Nebezpečí
|
Synonyms:
Cyclohexatriene
Filtered through a 0.2 µm filter.
AB208587.4000
AB208587.1000
ABCRAB208587.4000EA
0
CZK
ABCRAB208587.4000
ABCRAB208587.1000
Benzen ≥99.5% for HPLC
Benzen
Purity: 99.5%
CAS No.: 71-43-2
Molecular Formula: C6H6
Formula Weight: 78.11
Flash Point: -11°(12°F)
Physical Form: Liquid
Density: 0.874
Refractive Index: 1.5010
Odour: Aromatic
Melting Point: 5°
Boiling Point: 80°
Beilstein Reference: 969212
Merck Reference: 14,1066
UN No.: 1114
MDL No.: MFCD00003009
Vzorec:
C₆H₆ MW: 78.11 Bod varu: 80.1 Bod tání: 5.5 Hustota: 0.878 g/cm³ (20 °C) Bod vzplanutí: –11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003009 CAS číslo: 71-43-2 EINECS: 200-753-7 UN: 1114 ADR: 3 II REACH: 01-2119447106-44 Merck Index: 14,1066 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...