Tisk…

Cyclohexanemethylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (Cyclohexylmethyl)amine, (Aminomethyl)cyclohexane, Cyclohexylmethanamine

M-Clarity™ quality level = MQ100

101842-25G 101842-5G
SIAL101842-25GEA 4870 CZK
SIAL101842-25G SIAL101842-5G
Cyclohexanemethylamine, Sigma-Aldrich®
Cyclohexanemethylamine
Vzorec: C₇H₁₅N
Bod varu: 161…163 °C (1013 hPa)
Bod tání: –8 °C
Hustota: 0,871 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001520
CAS číslo: 3218-02-8
EINECS: 221-741-8
UN: 2735
ADR: 8,I

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR