Fenol Reag. Ph. Eur., ACS analytical reagent, Supelco®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
M-Clarity™ quality level = MQ100
1.00206.0250
1.00206.1000
1.00206.0250EA
2480
CZK
1.00206.0250
1.00206.1000
Fenol Reag. Ph. Eur., ACS analytical reagent, Supelco®
Fenol
For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues
Vzorec:
C₆H₅OH Bod varu: 181,7 °C (1013 hPa) Hustota: 1,0722 g/cm³ (20 °C) Bod vzplanutí: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS číslo: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1,II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Výsledky specifikačního testu
Solubility | 84 g/l (20 °C) |
Melting Point | 40.8 °C |
Molar Mass | 94.11 g/mol |
Bulk Density | 620 kg/m3 |
Boiling Point | 181.8 °C (1013 hPa) |
Vapor Pressure | 0.2 hPa (20 °C) |
Flash Point | 81 °C |
Explosion Limit | 1.3 - 9.5 %(V) |
Density | 1.06 g/cm3 (20 °C) |
pH | 5 (50 g/l, H2O, 20 °C) |
Ignition Point | 595 °C |
Hazard Class | 6.1 |
Hazard Class Description | Toxic materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...