Tisk…

Thiomorpholine ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15958.06 A15958.14 A15958.22
A15958.06EA 1890 CZK
A15958.06 A15958.14 A15958.22
Thiomorpholine ≥98%
Thiomorpholine

Purity: 98% CAS No.: 123-90-0 Molecular Formula: C4H9NS Formula Weight: 103.19 Flash Point: 67°(153°F) Refractive Index: 1.5384 Boiling Point: 175-177° Solubility: Miscible with water Beilstein Reference: 102550 Merck Reference: 14,9300 UN No.: 3267 MDL No.: MFCD00005974

Vzorec: C₄H₉NS
Bod varu: 175…177 °C
Hustota: 1,086
Bod vzplanutí: 67 °C (153 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005974
CAS číslo: 123-90-0
EINECS: 204-660-2
UN: 3267
ADR: 8,III
Merck Index: 14,9300

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR