Tisk…

p-Anisidinium chloride 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: p-Anisidine hydrochloride, 4-Methoxyaniline hydrochloride

401300250 401301000 401300050
ACRO401300250EA 1640 CZK
ACRO401300250 ACRO401301000 ACRO401300050
p-Anisidinium chloride 99%
p-Anisidinium chloride
Vzorec: C₇H₁₀ClNO
MW: 159.62
Bod tání: 216…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00036388
CAS číslo: 20265-97-8
EINECS: 243-657-0
UN: 2811
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Appearance Off-white to pink to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 214 to 219 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR