Tisk…

2,4-Difluorophenylboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(2,4-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

632694-10G 632694-5G
SIAL632694-10GEA 0 CZK
SIAL632694-10G SIAL632694-5G
2,4-Difluorophenylboronic acid pinacol ester, Sigma-Aldrich®
2,4-Difluorophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₅BF₂O₂
MW: 240,06 g/mol
MDL Number: MFCD05663880
CAS číslo: 288101-48-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR