Tisk…

3-Nitrophenylboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4,4,5,5-Tetramethyl-2-(3-nitrophenyl)-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

632708-1G 632708-5G
SIAL632708-1GEA 2390 CZK
SIAL632708-1G SIAL632708-5G
3-Nitrophenylboronic acid pinacol ester, Sigma-Aldrich®
3-Nitrophenylboronic acid pinacol ester
Vzorec: C₁₂H₁₆BNO₄
MW: 249,07 g/mol
MDL Number: MFCD05663879
CAS číslo: 68716-48-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR