Tisk…

2-Nitrophenylboronic acid pinacol ester ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2-Nitrophenylboronic acid Pinacol ester, 2-Nitrobenzeneboronic acid pinacol ester, 2-Nitrophenylboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

L19963.03 L19963.06
L19963.03EA 2280 CZK
L19963.03 L19963.06
2-Nitrophenylboronic acid pinacol ester ≥98%
2-Nitrophenylboronic acid pinacol ester

Purity: 98+% CAS No.: 190788-59-1 Molecular Formula: C12H16BNO4 Formula Weight: 249.08 Melting Point: 42-44° Beilstein Reference: 7708824

Vzorec: C₁₂H₁₆BNO₄
MW: 249.07
Bod tání: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD02179447
CAS číslo: 190788-59-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR