3-Aminophenylboronic acid pinacol ester 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester
H28448.06
H28448.03
H28448.06EA
8100
CZK
H28448.06
H28448.03
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester
Purity: 97%
CAS No.: 210907-84-9
Molecular Formula: C12H18BNO2
Formula Weight: 219.09
Melting Point: 87-93°
MDL No.: MFCD03453668
Vzorec:
C₁₂H₁₈BNO₂ MW: 219,09 g/mol Bod tání: 87…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453668 CAS číslo: 210907-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...