Ethyl isothiocyanatoformate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
196126-25G
196126-5G
SIAL196126-25GEA
6940
CZK
SIAL196126-25G
SIAL196126-5G
Ethyl isothiocyanatoformate, Sigma-Aldrich®
Ethyl isothiocyanatoformate
Vzorec:
C₄H₅NO₂S |
MDL Number:
MFCD00004814 CAS číslo: 16182-04-0 EINECS: 240-318-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...