Tisk…

Methyl isothiocyanatoacetate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L11046.03 L11046.06
L11046.03EA 1310 CZK
L11046.03 L11046.06
Methyl isothiocyanatoacetate ≥98%
Methyl isothiocyanatoacetate

Purity: 98% CAS No.: 21055-37-8 Molecular Formula: C4H5NO2S Formula Weight: 131.15 Storage Temperature: 2°C to 8°C Refractive Index: 1.5200 Odour: Irritating Boiling Point: 137-138°/35mm Beilstein Reference: 2074764 UN No.: 2922 MDL No.: MFCD00041119

Vzorec: C₄H₅NO₂S
Bod varu: 137…138 °C (35 mmHg)
Hustota: 1,210
MDL Number: MFCD00041119
CAS číslo: 21055-37-8
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR