1-Octylboronic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Octylboronic acid, 1-n-Octaneboronic acid, n-Octylboronic acid
L19964.03
L19964.06
L19964.14
L19964.03EA
886
CZK
L19964.03
L19964.06
L19964.14
1-Octylboronic acid ≥97%
1-Octylboronic acid
Purity: 97%
CAS No.: 28741-08-4
Molecular Formula: CH3(CH2)7B(OH)2
Formula Weight: 158.05
Melting Point: 81-85°
Beilstein Reference: 1739759
Vzorec:
C₈H₁₉BO₂ MW: 158.05 Bod tání: 81…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD01074560 CAS číslo: 28741-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...