Ethylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L19959.03
L19959.06
L19959.14
L19959.03EA
1560
CZK
L19959.03
L19959.06
L19959.14
Ethylboronic acid ≥98%
Ethylboronic acid
Purity: 98%
CAS No.: 4433-63-0
Molecular Formula: C2H7BO2
Formula Weight: 73.89
Storage Temperature: 2°C to 8°C
Odour: Odourless
Beilstein Reference: 1731546
MDL No.: MFCD01074536
Vzorec:
C₂H₇BO₂ Bod tání: 84…88 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074536 CAS číslo: 4433-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...