Tisk…

1-Decylboronic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Decylboronic acid

L19957.03 L19957.06 L19957.14
L19957.03EA 1180 CZK
L19957.03 L19957.06 L19957.14
1-Decylboronic acid ≥98%
1-Decylboronic acid

Purity: 98% CAS No.: 24464-63-9 Molecular Formula: CH3(CH2)9B(OH)2 Formula Weight: 186.10 Melting Point: 86-89° Beilstein Reference: 1743934

Vzorec: C₁₀H₂₃BO₂
MW: 186.1
Bod tání: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD01074638
CAS číslo: 24464-63-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR