1-Butylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Butylboronic acid, n-Butylboronic acid
M-Clarity™ quality level = MQ100
163244-1G
163244-25G
163244-5G
SIAL163244-1GEA
1130
CZK
SIAL163244-1G
SIAL163244-25G
SIAL163244-5G
1-Butylboronic acid, Sigma-Aldrich®
1-Butylboronic acid
Vzorec:
C₄H₁₁BO₂ MW: 101.94 |
MDL Number:
MFCD00002106 CAS číslo: 4426-47-5 EINECS: 224-607-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...