4-Benzyloxy-3,5-dimethylbenzoic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L14536.06
L14536.03
L14536.06EA
7310
CZK
L14536.06
L14536.03
4-Benzyloxy-3,5-dimethylbenzoic acid ≥98%
4-Benzyloxy-3,5-dimethylbenzoic acid
Purity: 98+%
CAS No.: 97888-80-7
Molecular Formula: C16H16O3
Formula Weight: 256.30
Melting Point: 157-160°
Beilstein Reference: 5045861
Vzorec:
C₁₆H₁₆O₃ Bod tání: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272572 CAS číslo: 97888-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...