Tisk…

Diphenyl sulphoxide ≥98%

Dodavatel: Thermo Scientific

Synonyms: Phenyl sulphoxide, sulphinyldibenzene

B22135.14 B22135.22 B22135.36
B22135.14EA 1880 CZK
B22135.14 B22135.22 B22135.36
Diphenyl sulphoxide ≥98%
Diphenyl sulphoxide

Purity: 98+% CAS No.: 945-51-7 Molecular Formula: C12H10OS Formula Weight: 202.28 Melting Point: 69-72° Boiling Point: 206-208°/13mm MDL No.: MFCD00002085

Vzorec: C₁₂H₁₀OS
MW: 202,28 g/mol
Bod varu: 206…208 °C (13 mmHg)
Bod tání: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002085
CAS číslo: 945-51-7
EINECS: 213-415-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR