Tisk…

Diglycolic anhydride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diglycolicanhydride

M-Clarity™ quality level = MQ200

D103705-100G D103705-25G
SIALD103705-100GEA 16450 CZK
SIALD103705-100G SIALD103705-25G
Diglycolic anhydride, Sigma-Aldrich®
Diglycolic anhydride
Vzorec: C₄H₄O₄
MW: 116.07
MDL Number: MFCD00006677
CAS číslo: 4480-83-5
EINECS: 224-761-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR