Tisk…

Diglycolic anhydride ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Diglycolicanhydride

B24498.09 B24498.18
B24498.09EA 1660 CZK
B24498.09 B24498.18
Diglycolic anhydride ≥97%
Diglycolic anhydride

Purity: 97% CAS No.: 4480-83-5 Molecular Formula: C4H4O4 Formula Weight: 116.07 Melting Point: 91-94° Boiling Point: 240-241° MDL No.: MFCD00006677

Vzorec: C₄H₄O₄
Bod varu: 240…241 °C
Bod tání: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00006677
CAS číslo: 4480-83-5
EINECS: 224-761-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR