Tisk…

3-(Benzyloxy)benzoic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Benzyloxybenzoic acid

H32882.03 H32882.06 H32882.14
H32882.03EA 1130 CZK
H32882.03 H32882.06 H32882.14
3-(Benzyloxy)benzoic acid ≥98%
3-(Benzyloxy)benzoic acid

Purity: 98% CAS No.: 69026-14-8 Molecular Formula: C14H12O3 Formula Weight: 228.25 Melting Point: 133-135° MDL No.: MFCD00527901

Vzorec: C₁₄H₁₂O₃
MW: 228,25 g/mol
Bod tání: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00527901
CAS číslo: 69026-14-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR