2-(Benzyloxy)benzoic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Benzyloxybenzoic acid
A12425.09
A12425.18
A12425.30
A12425.09EA
2030
CZK
A12425.09
A12425.18
A12425.30
2-(Benzyloxy)benzoic acid ≥98%
2-(Benzyloxy)benzoic acid
Purity: 98%
CAS No.: 14389-86-7
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 76-78°
MDL No.: MFCD00051940
Vzorec:
C₁₄H₁₂O₃ MW: 228,25 g/mol Bod tání: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051940 CAS číslo: 14389-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...