Tisk…

4'-(Trifluoromethyl)biphenyl-2-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid

M-Clarity™ quality level = MQ100

346357-5G 346357-1G
SIAL346357-5GEA 1390 CZK
SIAL346357-5G SIAL346357-1G
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid, Sigma-Aldrich®
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
Vzorec: C₁₄H₉F₃O₂
MW: 266,22 g/mol
MDL Number: MFCD00075353
CAS číslo: 84392-17-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR