2,3-Dihydrofuran, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
200018-100ML
200018-1L
SIAL200018-100MLEA
1960
CZK
SIAL200018-100ML
SIAL200018-1L
2,3-Dihydrofuran, Sigma-Aldrich®
2,3-Dihydrofuran
Vzorec:
C₄H₆O MW: 70,09 g/mol |
MDL Number:
MFCD00003205 CAS číslo: 1191-99-7 EINECS: 214-747-7 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...