1-Methyl-2-phenylindole ≥99%
Dodavatel: Thermo Fisher Scientific
B22105.09
B22105.18
B22105.30
B22105.09EA
968
CZK
B22105.09
B22105.18
B22105.30
1-Methyl-2-phenylindole ≥99%
1-Methyl-2-phenylindole
Purity: 99%
CAS No.: 3558-24-5
Molecular Formula: C15H13N
Formula Weight: 207.28
Melting Point: 98-99°
Beilstein Reference: 148770
Vzorec:
C₁₅H₁₃N MW: 207.27 Bod tání: 98…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022892 CAS číslo: 3558-24-5 EINECS: 222-618-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...