Tisk…

α-Ionone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (E)-4-(2,6,6-Trimethylcyclohex-2-en-1-yl)but-3-en-2-one

M-Clarity™ quality level = MQ300

W259403-100G-K W259403-1KG-K W259403-5KG-K
SIALW259403-100G-KEA 2850 CZK
SIALW259403-100G-K SIALW259403-1KG-K SIALW259403-5KG-K
α-Ionone, Sigma-Aldrich®
α-Ionone
Vzorec: C₁₃H₂₀O
MW: 192,3 g/mol
MDL Number: MFCD00001565
CAS číslo: 127-41-3
EINECS: 204-841-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR