Tisk…

(E)-β-Ionone (synthetic) 96%

Dodavatel: Thermo Scientific

Synonyms: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone

297130050 297131000 297135000
ACRO297130050EA 751 CZK
ACRO297130050 ACRO297131000 ACRO297135000
(E)-β-Ionone (synthetic) 96%
(E)-β-Ionone
Vzorec: C₁₃H₂₀O
MW: 192,30 g/mol
Bod vzplanutí: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00001549
CAS číslo: 14901-07-6
EINECS: 201-224-3
UN: 3082

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5215 (20°C, 589 nm)
GC ≥95 %
Specific gravity (25°C) 0.941 to 0.947

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR