Tisk…

(R)-(+)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

M-Clarity™ quality level = MQ100

237442-1G 237442-5G
SIAL237442-1GEA 1770 CZK
SIAL237442-1G SIAL237442-5G
(R)-(+)-1-(1-Naphthyl)ethylamine, Sigma-Aldrich®
(R)-(+)-1-(1-Naphthyl)ethylamine
Vzorec: C₁₂H₁₃N
MDL Number: MFCD00064114
CAS číslo: 3886-70-2
EINECS: 223-425-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR