(R)-(+)-1-(1-Naphthyl)ethylamine 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
A16918.03
A16918.06
A16918.14
A16918.03EA
1410
CZK
A16918.03
A16918.06
A16918.14
(R)-(+)-1-(1-Naphthyl)ethylamine 99%
(R)-(+)-1-(1-Naphthyl)ethylamine
Purity: 99%
CAS No.: 3886-70-2
Molecular Formula: C12H13N
Formula Weight: 171.24
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₁₂H₁₃N MW: 171.24 Bod varu: 152…153 °C (11 mmHg) Hustota: 1.060 Bod vzplanutí: >110 °C (230 °F) |
MDL Number:
MFCD00064114 CAS číslo: 3886-70-2 EINECS: 223-425-5 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...