Diethyl chloromalonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Diethyl 2-chloromalonate
M-Clarity™ quality level = MQ100
437786-25ML
437786-5ML
SIAL437786-25MLEA
6670
CZK
SIAL437786-25ML
SIAL437786-5ML
Diethyl chloromalonate, Sigma-Aldrich®
Diethyl chloromalonate
Vzorec:
C₇H₁₁ClO₄ |
MDL Number:
MFCD00009140 CAS číslo: 14064-10-9 EINECS: 237-913-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...