Dimethyl bromomalonate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
359890-25ML
359890-5ML
SIAL359890-25MLEA
5250
CZK
SIAL359890-25ML
SIAL359890-5ML
Dimethyl bromomalonate, Sigma-Aldrich®
Dimethyl bromomalonate
Vzorec:
C₅H₇BrO₄ |
MDL Number:
MFCD00025865 CAS číslo: 868-26-8 EINECS: 212-775-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...