Tisk…

(R)-(-)-2-Hydroxyisovaleric acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid

55452-1G 55452-5G
SIAL55452-1GEA 4950 CZK
SIAL55452-1G SIAL55452-5G
(R)-(-)-2-Hydroxyisovaleric acid, Sigma-Aldrich®
(R)-(-)-2-Hydroxyisovaleric acid
Vzorec: C₅H₁₀O₃
MW: 118,13 g/mol
MDL Number: MFCD00066442
CAS číslo: 17407-56-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR