1-Butyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®
Dodavatel: Merck
55292-1KG-F
55292-100G-F
SIAL55292-1KG-FEA
10080
CZK
SIAL55292-1KG-F
SIAL55292-100G-F
1-Butyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtetrachloroaluminate
Vzorec:
C₈H₁₅AlCl₄N₂ MW: 308,01 g/mol |
CAS číslo:
80432-09-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...