Adenine sulphate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
6-Aminopurine hemisulphate salt, 7H-Purin-6-amine hemisulphate, Adenine hemisulphate salt
A9126-100G
A9126-25G
A9126-1KG
SIALA9126-100GEA
14010
CZK
SIALA9126-100G
SIALA9126-25G
SIALA9126-1KG
Adenine sulphate, Sigma-Aldrich®
Adenine sulphate
Vzorec:
C₅H₅N₅·½H₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00213655 CAS číslo: 321-30-2 EINECS: 206-286-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...