Tisk…

2-(4-Fluorophenyl)indole ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A13255.03 A13255.06
A13255.03EA 851 CZK
A13255.03 A13255.06
2-(4-Fluorophenyl)indole ≥99%
2-(4-Fluorophenyl)indole

Purity: 99% CAS No.: 782-17-2 Molecular Formula: C14H10FN Formula Weight: 211.24 Odour: Odourless Melting Point: 186-191° Beilstein Reference: 744766 MDL No.: MFCD00068178

Vzorec: C₁₄H₁₀FN
MW: 211.24
Bod tání: 186…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00068178
CAS číslo: 782-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR