Tisk…

Indol-3-ylacetonitrile, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Indole-3-Acetonitrile, 3-Indolylacetonitrile, 2-(1H-Indol-3-yl)acetonitrile, 3-Indoleacetonitrile

M-Clarity™ quality level = MQ100

129453-25G 129453-5G
SIAL129453-25GEA 4660 CZK
SIAL129453-25G SIAL129453-5G
Indol-3-ylacetonitrile, Sigma-Aldrich®
Indol-3-ylacetonitrile
Vzorec: C₁₀H₈N₂
MW: 156,19 g/mol
MDL Number: MFCD00005628
CAS číslo: 771-51-7
EINECS: 212-232-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR