Tisk…

Methyl indole-5-carboxylate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 1H-indole-5-carboxylate

M-Clarity™ quality level = MQ100

511188-1G 511188-5G
SIAL511188-1GEA 1250 CZK
SIAL511188-1G SIAL511188-5G
Methyl indole-5-carboxylate, Sigma-Aldrich®
Methyl indole-5-carboxylate
Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
MDL Number: MFCD00153023
CAS číslo: 1011-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR