Tisk…

1-Boc-indole ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: tert-Butyl 1-indolecarboxylate

L17601.03 L17601.06
L17601.03EA 1690 CZK
L17601.03 L17601.06
1-Boc-indole ≥97%
1-Boc-indole

Purity: 97% CAS No.: 75400-67-8 Molecular Formula: C13H15NO2 Formula Weight: 217.27 Storage Temperature: 2°C to 8°C Refractive Index: 1.5430 Boiling Point: 113-114°/0.2mm Beilstein Reference: 4674239 MDL No.: MFCD02093939

Vzorec: C₁₃H₁₅NO₂
Bod varu: 113…114 °C (0,2 mmHg)
Hustota: 1,070
MDL Number: MFCD02093939
CAS číslo: 75400-67-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR