Tisk…

1-Benzyl-4-piperidone ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone

A12390.22 A12390.14 A12390.36
A12390.22EA 2950 CZK
A12390.22 A12390.14 A12390.36
1-Benzyl-4-piperidone ≥98%
1-Benzyl-4-piperidone

Purity: 98+% CAS No.: 3612-20-2 Molecular Formula: C12H15NO Formula Weight: 189.26 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5399 Boiling Point: 248° Beilstein Reference: 128556 MDL No.: MFCD00006192

Vzorec: C₁₂H₁₅NO
MW: 189,26 g/mol
Bod varu: 248 °C
Hustota: 1,059
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD00006192
CAS číslo: 3612-20-2
EINECS: 222-782-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR