Tisk…

1-Boc-piperazine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

M-Clarity™ quality level = MQ200

343536-1G 343536-25G 343536-5G
SIAL343536-1GEA 917 CZK
SIAL343536-1G SIAL343536-25G SIAL343536-5G
1-Boc-piperazine, Sigma-Aldrich®
1-Boc-piperazine
Vzorec: C₉H₁₈N₂O₂
MW: 186.25
MDL Number: MFCD00075265
CAS číslo: 57260-71-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR