Tisk…

Nerol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

268909-100ML 268909-5ML
SIAL268909-100MLEA 1520 CZK
SIAL268909-100ML SIAL268909-5ML
Nerol, Sigma-Aldrich®
Nerol
Vzorec: C₁₀H₁₈O
MDL Number: MFCD00063204
CAS číslo: 106-25-2
EINECS: 203-378-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR