Nerol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
cis-3,7-Dimethyl-2,6-octadien-1-ol
M-Clarity™ quality level = MQ300
W277002-1KG-K
W277002-4KG-K
W277002-9KG-K
SIALW277002-1KG-KEA
3070
CZK
SIALW277002-1KG-K
SIALW277002-4KG-K
SIALW277002-9KG-K
Nerol, Sigma-Aldrich®
Nerol
Vzorec:
C₁₀H₁₈O MW: 154,25 g/mol |
MDL Number:
MFCD00063204 CAS číslo: 106-25-2 EINECS: 203-378-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...